Brady™ “Danger: Hazardous Waste Storage Area…” Signs

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

Manufacturer: Brady™ 70375

 View more versions of this product

Catalog No. 19-016660

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.,10 in.
    Height (Metric) 25.4cm,25.4cm
    Material Premium Fiberglass
    Certifications/Compliance ASTM D 1593, ASTM D 638, ASTM D 638, ASTM D 646
    Message Danger Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Quantity Each
    Style Surface Mount
    Temperature Range (English) -40 to 193°F,-40 to 193°F
    Temperature Range (Metric) -40 to 90°C,-40 to 90°C
    Tensile Strength 9000psi
    Thickness 0.1 in.,0.1 in.
    Color Black, Red on White,Black, Red on White
    Type Admittance sign
    Width (English) 14 in.,14 in.
    Width (Metric) 35.5cm,35.5cm
    For Use With (Application) Industrial, utility, commercial and institutional environment

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit