Brady™ “Danger: Hydrogen Flammable Gas No Smoking or Open Flames” Signs Available on GSA/VA Contract for Federal Government customers only.

Used to identify equipment, rooms and materials which contain or are contaminated with hazardous agents

Manufacturer: Brady™ 25453

 View more versions of this product

Catalog No. 19-036180

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.
    Height (Metric) 25.4cm
    Material Plastic
    Certifications/Compliance OSHA
    Message Danger Hydrogen Flammable Gas No Smoking or Open Flames
    Quantity Each
    Temperature Range (Metric) -40 to 80°C
    Thickness 0.06 in.,0.06 in.
    Width (English) 14 in.
    Width (Metric) 35.5cm
    For Use With (Application) Industrial, utility, commercial and institutional environment
    Temperature Range (English) -40 to 175°F
    Color Black, Red on White,Black, Red on White
    Type Chemical and hazardous materials sign

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit