Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Danger: Keep Off” Signs Available on GSA/VA Contract for Federal Government customers only.

Identify areas that certain, items or action are restricted

Manufacturer:  Brady™ 22110

 View more versions of this product

Catalog No. 19-035-601


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 1910.145, ASTM D 1593
Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs and equipment warning signs, recommended for both indoor and outdoor use
25.4cm
Danger Keep Off
Surface Mount
-40 to 80°C
Admittance sign
35.5cm
Black, Red on White
10 in.
Plastic
Each
-40 to 175°F
0.06 in.
14 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit