Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Danger: Keep Off” Signs Available on GSA/VA Contract for Federal Government customers only.

Identify areas that certain, items or action are restricted

Manufacturer:  Brady™ 84091

 View more versions of this product

Catalog No. 19-035-602


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 1910.145, PSTC-33
Used for safety signs, identification of fire alarms, extinguishers and emergency exits
17.7cm
Danger Keep Off
Surface Mount
-40 to 110°C
Admittance sign
25.4cm
Black, Red on White
7 in.
Polyester
Each
-40 to 230°F
0.006 to 0.01 in.
10 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit