Brady™ “Danger: Pesticide Storage…” Signs Available on GSA/VA Contract for Federal Government customers only.

For identifying equipment, rooms and materials that contain or are contaminated with hazardous agents

Manufacturer: Brady™ 70479

 View more versions of this product

Catalog No. 19-035720

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Height (English) 10 in.,10 in.
    Temperature Range (English) -40 to 193°F,-40 to 193°F
    Temperature Range (Metric) -40 to 90°C,-40 to 90°C
    Width (English) 14 in.,14 in.
    Height (Metric) 25.4cm,25.4cm
    Material Premium Fiberglass
    Certifications/Compliance ANSI Z535, OSHA 1910.145, ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646, ASTM D 635
    Message Danger Pesticide Storage Authorized Personnel Only
    Quantity Each
    Style Surface Mount
    Tensile Strength 9000psi
    Thickness 0.1 in.,0.1 in.
    Color Black, Red on White,Black, Red on White
    Type Admittance sign
    Width (Metric) 35.56cm,35.56cm
    For Use With (Application) Industrial, utility, commercial and institutional environment

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit