Brady™ “Danger: Wear Goggles And Rubber Gloves” Signs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Brady™ 22349

 View more versions of this product

Catalog No. 19-102415

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D 1593, ANSI Z535, OSHA 1910.145,ASTM D 1593, ANSI Z535, OSHA 1910.145
    Message Danger Wear Goggles And Rubber Gloves When Handling Chemicals
    Quantity Each
    Style Surface Mount
    Type Chemical and hazardous materials sign
    For Use With (Application) Light duty industrial, utility, commercial and institutional environments for both indoor and outdoor use
    Height (English) 7 in.,7 in.
    Material Plastic
    Temperature Range (English) -40 to 175°F
    Temperature Range (Metric) -40 to 80°C
    Thickness 0.06 in.
    Color Black, Red on White,Black, Red on White
    Width (English) 10 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit