Brady™ Electrical Hazard Signs: WARNING - HIGH VOLTAGE Available on GSA/VA Contract for Federal Government customers only.

Identify electrical hazard and high voltage areas

Manufacturer: Brady™ 84942

 View more versions of this product

Catalog No. 19-035-921

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.,10 in.
    Height (Metric) 25.40cm,25.40cm
    Thickness 0.006 to 0.01 in.
    Width (English) 14 in.
    Style Surface Mount
    Material Polyester
    Certifications/Compliance ANSI Z535, OSHA 1910.145, PSTC-33,,ANSI Z535, OSHA 1910.145, PSTC-33,
    Message Warning High Voltage
    Quantity Each
    Temperature Range (English) -40 to 230°F
    Temperature Range (Metric) -40 to 110°C
    Color Black on Red,Black on Red
    Type Electrical hazard sign
    Width (Metric) 35.56cm
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit