Brady™ ENV™ - MAXX Enhanced Oil Only Sorbent Pads

Oil only sorbent equally effective on land or water

Manufacturer: Brady™ ENV300

 View more versions of this product

Catalog No. 19-000-160

  Check Availability
Add to Cart

    Specifications

    Product Type ENV MAXX Enhanced Sorbent Pad
    For Use With (Application) Overspray Leaks and Drips, Workstations, Drums - Liquid Storage, Outfalls and Discharge Ponds
    Weight Capacity Medium
    Capacity 28 gal./bale
    Dimensions (L x W) 38.1 x 48.2cm (15 x 19 in.)
    Quantity 100/bale
    Color White

    • Single-ply construction
    • Repels water and floats indefinitely even when saturated
    • For use with oil and petroleum-based fluids
    • Available in two sizes

    MAXX refers to enhanced performance without extra weight

    Applications:

    Marine applications, over spray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds, emergency spill response.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit