Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Flip Style Pressure Vehicle Placards, one piece frame
For easy identification of chemical and hazardous materials. Brady Flip Style, Pressure Vehicle Placards, made of durable, heavy-duty aluminum and assures stability in the wind
Supplier: Brady™ 63213
Includes: Legend Set 1:
Blank, Flammable Gas 2, Non-Flammable Gas 2, Oxygen 2, Inhalation Hazard 2
Description
- Flipping numerals gives flexibility to set-up any 4-digit identification number in seconds
- Durable, heavy-duty aluminum frame assures stability in the wind
Specifications
Multi-colored | |
34.59 x 34.59cm | |
Aluminum | |
13.62 in. | |
13.62 in. |
Diamond | |
Legend labels set 1 | |
1 Ea. | |
34.59 mm | |
34.59 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction