Learn More
Brady™ FreezerBondz II Rectangular Labels
Freezer-safe labels for Brady printers, including the IDXPERT and LABXPERT
Manufacturer:  Brady™ X130492X
 View more versions of this product
Catalog No. 22-500-785
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Acrylic,Permanent
RoHS, ASTM D 1000
Semi-Gloss
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
0.93cm
-320°F
0.0032 in.
Cartridge, Label
Cryogenic Label
Black,White
Surface Mount
0.37 in.
iDXPERT™, LabXpert™ printer
Cartridges
-196°C
0.8 in.
Polyester, FreezerBondz™ II
275 Pack