Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Freezerbondz Polyester (B-490) LABXPERT™ Labels

Identification label for frozen surfaces including glass and polypropylene stored in liquid nitrogen

Manufacturer:  Brady™ X125490X

 View more versions of this product

Catalog No. 22-500-746

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

  • Smooth surface label made of Freezerbondz Polyester
  • Moisture resistant label with cold-weather performance
  • Original Freezerbondz I label withstands extremely low temperatures
  • For LABEXPERT™ printer (also fits in IDXPERT™ printer)
  • Matte finish, black on white label
  • Environment: Autoclave, Freezer, Hot Water Baths (100°C), Liquid Nitrogen
  • Available in different sizes and printable areas
  • Printer compatibility: LABXPERT

Intended use of product is out of scope of the RoHS Directive

Description & Specifications

Specifications

Smooth
Matte
LABXPERT Printer
25.4 x 44.45mm
225/Cartridge
Black,White
Cryo Vial, Laboratory Identification
1 x 1.750 in.
FreezerBondz™ Polyester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit