Learn More
Brady™ GlobalMark™ Industrial Label Makers
Versatile industrial-grade label makers create custom labels quickly and simply
Manufacturer:  Brady™ 76801
Catalog No. 19-201-064
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Keyboard, power cord, cleaning kit, USB cable and driver diskette, user's guide, and tutorial. ValuePLUS Package also includes the MarkWare Facility Identification Software, one roll of 4 in. white vinyl tape, and one roll of black ribbon.
Description
Description
Create signs and labels, including vinyl lettering, magnetic backed, static cling, and glow-in-the-dark, from 0.5 to 4 in. wide; make larger signs by tiling sections together.
  • 290 built-in symbols
  • Hundreds of built-in templates in six industrial applications (signs, pipe markers, Right To Know, tags, equipment I.D., and warehouse I.D.)
  • Choice of three languages (English, Spanish and French)
  • Available in three models
GlobalMark Color and Cut Model
  • Prints in single color, multiple spot colors, or blended colors Includes built-in X-Y axis plotter/cutter for cutting out images and letters “Tape Saver” feature prints “multiple-up” labels
GlobalMark Multicolor Model Offers the same print options as the Color and Cut Model without the built-in cutter GlobalMark Monocolor Model
    Has a monochrome touchscreen and prints one color only on one color tape Does not include built-in cutter
Warranty and Services
Six months for all models except ValuePLUS, which is 18 months
Specifications
Specifications
Arc Flash labeling, Barcode labeling, Data communications labeling, Facility, Safety and maintenance identification, Laboratory identification, Lean or 5S labeling, Lockout tagout labeling, Panel identification, Pipe and valve marking, Product identification, Security identification
16 MB
300 dpi
13.607 kg
1 Ea.
LED
Keyboard, power cord, cleaning kit, USB cable and Brady file management utility, 290 built-in symbols, built-in X-Y axis plotter/cutter for cutting out images and letters, tape saver feature prints multiple-up labels
AC power supply and USB, serial connectivity
30 lb.
5 Years
Tape
SDS
Documents
Accessories