Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ Reflective Tapes

Shines back at viewer brightly when light shines on label. Brady GlobalMark Reflective Tapes have permanent adhesive/outdoor grade.

$262.00 - $998.00

Specifications

Format Tape
Product Type Labeling Tape
Quantity Each
For Use With (Application) Outdoor
For Use With (Equipment) GlobalMark
View More Specs

Products 9
Catalog Number Mfr. No. Width (English) Length (English) Color Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Length (English) Color Price Quantity & Availability  
19-201-196
View Documents
Brady™
76705
4 in. 50 ft. Silver Each for $998.00
Add to cart
 
19-201-199
View Documents
Brady™
76706
2.25 in. 50 ft. Silver Each for $498.50
Add to cart
 
19-201-202
View Documents
Brady™
76707
1.12 in. 50 ft. Silver Each for $262.00
Add to cart
 
19-108-042
View Documents
Brady™
99221
1.375 in. 50 ft. Orange Each for $262.00
Add to cart
 
19-108-041
View Documents
Brady™
99220
2.25 in. 50 ft. Orange Each for $498.50
Add to cart
 
19-108-029
View Documents
Brady™
96060
4 in. 50 ft. Yellow Each for $998.00
Add to cart
 
19-108-040
View Documents
Brady™
99219
4 in. 50 ft. Orange Each for $998.00
Add to cart
 
19-108-031
View Documents
Brady™
96062
1.375 in. 50 ft. Yellow Each for $262.00
Add to cart
 
19-108-030
View Documents
Brady™
96061
2.25 in. 50 ft. Yellow Each for $498.50
Add to cart
 
Description

Description

  • Reflects when light shines on label and shines brightly
  • Not recommened for plotter cutting
  • Available in two colors, yellow and orange
Specifications

Specifications

Tape
Each
GlobalMark
Surface Mount
Labeling Tape
Outdoor
Vinyl
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit