Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ IDXPERT™ and LABXPERT™ FreezerBondz II Labels: Rectangular with Circles Available on GSA/VA Contract for Federal Government customers only.

Labels withstand environments from LN2 to autoclave, helping preserve research while maintaining sample traceability

$190.30

Specifications

Adhesive Type Acrylic, Permanent
Color Black, White
Product Type Label
Finish Semi-Gloss
For Use With (Application) Centrifuge tube, Eppendorf tube, Laboratory identification, Tube, Vial, Vial top
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (English) Quantity Height (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Quantity Height (English) Width (English) Price Quantity & Availability  
22-500-532 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Brady™
X118492
0.37 in. 300/Pk. 0.37 in. 1 in.
Each for $190.30
Add to cart
 
22-500-531
View Documents
Brady™
X117492
0.44 in. 300/Pk. 0.5 in. 1 in.
Each for $190.30
Add to cart
 
22-500-533 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Brady™
X120492
0.37 in. 300/Pk. 0.5 in. 1 in.
Each for $190.30
Add to cart
 
Description

Description

Labels withstand environments from LN2 to autoclave, helping preserve research while maintaining sample traceability

  • Made of polyester with permanent acrylic adhesive
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides viewing window for liquid level inside vial
  • Survives exposure to common laboratory environments, such as liquid nitrogen, freezers down to -196°C (-320.8°F), autoclaves, and water baths up to 121°C (249.8°F)
  • Label sizes available for vials, slides, well plates, straws, petri dishes, boxes, and general applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

For use with Brady LABXPERT and IDXPERT printers

Specifications

Specifications

Acrylic, Permanent
Label
Centrifuge tube, Eppendorf tube, Laboratory identification, Tube, Vial, Vial top
Surface Mount
Black, White
Semi-Gloss
Polyester
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit