Learn More
Brady™ IDXPERT™ ToughBond™ Label Tapes
IDXPERT™ ToughBond™ Label Tapes are thermal transfer printable, glossy Permanent Polyester (B-422) labels that are ideal for use in all commercial and industrial applications.
Manufacturer:  Brady™ XC375422
 View more versions of this product
Catalog No. 19-078-746
Add to cart
Description
Description
  • Aggressive adhesive good for powder-coated and textured flat surfaces including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • All-in-one cartridge includes ribbon and label supply
Specifications
Specifications
Non-sterile
Acrylic, Permanent
UL, UL969, CSA,C22.2 0.15-95, RoHS, Pass 2002/95/EC
9.53 mm
0.357 in.
iDXPERT™
1
Continuous
-40°C to +120°C
0.102 mm
9.53 mm
Label Tape
Safety
Black, White
Surface Mount
Gloss
Electronic PCB and component identification, bar code label and rating plates and solar panel identification
0.9 cm
9.14 m
-40 to 248°F
0.004 in.
19 in.
Polyester
1 Roll(s)