Learn More
Brady™ Inspection and Material Control Tags
Manufacturer:  Brady™ 86568
 View more versions of this product
Catalog No. 19-040-432
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OSHA
0.37 in.
3 to 5 Years
5.75 in.
10 nylon ties
-40° to 100°C
3 in.
Polyester
Red on White
0.93cm
For marking on-the-spot warning of dangerous conditions
14.6cm
-40° to 212°F
0.01 in.
7.6cm
Quality Control Date/By