Learn More
Brady™ IP™ Thermal Transfer Printer Ribbons
Have strong environmental resistance against smearing and chemicals
Manufacturer:  Brady™ IPR4302X
 View more versions of this product
Catalog No. 22-500-840
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Use with Brady IP Thermal Transfer Printers
  • Sharp-edge print quality to maximize bar code readability
  • Resistant against smearing and chemicals
  • Ribbons include spindle
Description & Specifications
Specifications
Black
2.54cm
Thermal transfer printer, IP300, IP600
984 ft.
2.36 in.
Ribbon
Each
RoHS 2005/618/EC MCV, 2002/95/EC
Smearing and chemicals Resistant
Spindle
299.9m
1 in.
Resin,Wax