Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Key Retaining Keyed Different Steel Padlocks with 0.75 inch Shackle Locks
Key retaining safety feature prevents key from being removed until closed
Supplier: Brady™ 123281
View more versions of this product
Safety Services Available
Overview
LOTO Assessments
Includes: 1 key
Description
- Key-retaining steel padlock has a 0.75 in. reinforced laminated steel shackle
- Hardened steel body withstands severe physical abuse and tampering
- Offers individually coated body plates that provide superior rust protection and corrosion resistance
- Available in a variety of different colors
Specifications
Purple |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction