Learn More
Brady™ Lab Pal™ Printer Labels
Can be used in a wide range of temperatures—from autoclaves to freezers
Manufacturer:  Brady™ PAL375488/PRC
Catalog No. 11-877-177
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Durable, chemical-, and solvent-resistant labels for use with portable LabPal printer
  • Oversized liner allows easy label removal
  • White matte polyester and white matte nylon cloth are ideal for vial identification
  • White matte polyester recommended for use in only freezers and autoclaves; xylene-resistant and ideal for slide identification
Order Info
Roll length: 21 ft. (6.4m). Additional labels available. For ordering information, contact your Customer Service Representative or visit www.fishersci.com.