Learn More
Brady™ LABPALl™ Label Printer
Create legible lab labels quickly
Manufacturer:  Brady™ LABPAL
Catalog No. 11-877-175
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Easy-to-use, affordable, handheld thermal transfer printer design makes labeling more efficient.
  • Prints durable, nonsmearing labels that withstand liquid nitrogen (-196°C) to autoclave and chemicals such as ethanol, IPA, xylene and DMSO
  • Sizes specifically made for vials, slides, and well plates
  • 0.6, 1.5, and 2.0mL vial sizes preprogrammed into printer to make labeling samples quick and simple
  • Features time/date stamp, banner mode, serialization for aliquotting, five font sizes, and special laboratory symbols
Order Info
Uses six “AA” batteries or AC power with optional adapter (sold separately). Weight (with batteries and quick “drop-in” tape cartridge): 1 1/4 lb. (0.5kg)
Compliance
FCC Class A approved.
Specifications
Specifications
Printer