Learn More
Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels
Use these labels for a variety of general labeling needs. Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels are ideal for LABXPERT™ and IDXPERT™ printers.
Manufacturer:  Brady™ X117499X
 View more versions of this product
Catalog No. 22-500-760
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Nylon cloth labels
  • Matte finish
  • Permanent acrylic adhesive
Description & Specifications
Specifications
Acrylic,Permanent
UL
Powder Coated, Smooth, Textured/Rough
Bottles, conical tube, general and industrial identification, general identification, inside-of-panel component marking, laboratory identification, outside-of-panel markings, panel identification, petri dish, tube
12.70mm
Rectangle and circle
Cartridge, Label
Printable Label
Black,White
11.18mm
Matte
iDXPERT™, LabXpert™ printer
300 Labels
25.40mm
Nylon Cloth