Learn More
Brady™ LABXPERT™ Printer Freezerbondz Labels
Excellent cold-weather performance
$120.30
Specifications
Finish Matte
Description Surface Mount
View More Specs
Products 6
Catalog Number Mfr. No. Color Height (English) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Height (English) Height (Metric) Price Quantity & Availability  
11-878-149
View Documents
Brady™
X82499ORBK
Black, Orange 0.3 in. 0.9 cm
Each for $120.30
Add to cart
11-878-150
View Documents
Brady™
X82499RDBK
Black, Red 0.3 in. 0.9 cm
Each for $120.30
Add to cart
11-878-151
View Documents
Brady™
X82499YLBK
Black, Yellow 0.3 in. 0.9 cm
Each for $120.30
Add to cart
11-878-152
View Documents
Brady™
X83499ORBK
Black, Orange 0.5 in. 1.2 cm
Each for $120.30
Add to cart
11-878-153
View Documents
Brady™
X83499RDBK
Black, Red 0.5 in. 1.2 cm
Each for $120.30
Add to cart
11-878-154
View Documents
Brady™
X83499YLBK
Black, Yellow 0.5 in. 1.2 cm
Each for $120.30
Add to cart
Description
Description
  • Made of Freezerbondz polyester or nylon cloth
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Ideal for autoclave, freezer, hot water baths (100°C), liquid nitrogen
  • For use with centrifuge tube, cryo vial, eppendorf tube, laboratory identification
  • Moisture resistance
  • Smooth surface with matte finish
  • Compatible with LABXPERT printer
Specifications
Specifications
Matte
Surface Mount