Learn More
Brady™ LABXPERT™ Printer Labels
Specialty laboratory labels designed for use with these specific label printers
Manufacturer:  Brady™ XSL86461X
 View more versions of this product
Catalog No. 22-500-796
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Printable, self-laminating high-performance laboratory identification for LABXPERT printer.
  • Polyester label withstands temps -196° to +121°C
Ultrathin construction prevents jamming in racks & centrifuge
Description & Specifications
Specifications
Acrylic,Permanent
Surface Mount
0.6 in.
LabXpert™ printer
-320.8 to 249.8°F
2.62 in.
Polyester
150 Roll(s)
Black,Translucent,White
Matte
Centrifuge tube, cryo vial, Eppendorf tube, laboratory identification, tube, vial
1.52cm
-196 to 121°C
Cartridge, Label
Laboratory Label