Learn More
Brady™ Large Lockout Station w/Components and 12 Steel Padlocks, 25 Tags
Large Lockout Station withComponents and 12 Steel Padlocks, 25 Tags includes components needed to lock out most types of electrical control points.
Supplier:  Brady™ 99697
Catalog No. 19-000-055
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Centralizes lockout activity needs by organizing the tools, padlocks and padlock keys in one location
  • Designed with the durability required for an industrial workplace
Specifications
Specifications
Large Lockout Station
5 x 19.5 x 26 in.
(1) All Purpose Cable Lockout w/8 ft. Sheathed Cable, (1) Large Universal Valve Lockout w/ Blocking Arm, (1) 110V Plug Lockout, (1) 3-In-1 Plug Lockout, (1) 1/4 to 1 in. Ball Valve Lockout, (1) 1 to 2.5 in. Gate Valve Lockout, (1) 1 to 6 1/2 in. Adjustable Gate Valve Lockout, (2) Labeled Lockout Hasps, (12) Brady Steel Padlocks, (25) Lockout Tags
49.53 cm
Product Suggestions
Product Suggestions