Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Laser and Ink Jet Sign Blanks: Danger Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  Brady™ 12907

 View more versions of this product

Catalog No. 19-035455


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Acrylic,Pressure-sensitive
Danger
Used for safety, maintenance and general markings within a facility
12.7cm
Polyester
25 Pack
Black,Red,White
5 in.
Laser and inkjet printer
7 in.
Sign Blank
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit