Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “No Smoking In This Area” Signs

To reinforce policies that prohibit smoking and help keep employees and visitors safe

Manufacturer:  Brady™ 72091

 View more versions of this product

Catalog No. 19-016852


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Material: Premium Fiberglass
14 in.
193°F
No Smoking In This Area
-40°C
No Smoking Sign
50.8cm
White on Red
Each
Indoor and outdoor use, industrial, utility, commercial and institutional environment
35.5cm
90°C
-40°F
0.1 in.
20 in.
OSHA
Fiberglass
9000psi
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit