Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Padlock Stations

Constructed of 14 gauge steel with red plastic coating

$30.80 - $46.90

Specifications

Depth (Metric) 3.1cm
View More Specs

Products 2
Catalog Number Mfr. No. Product Type For Use With (Application) Height (Metric) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type For Use With (Application) Height (Metric) Width (Metric) Price Quantity & Availability  
19-103-591
View Documents
Brady™
LR060E
Small Padlock Station Holds up to 4 padlocks 5.7cm 7.6cm
Each for $30.80
Add to cart
 
19-103-592
View Documents
Brady™
LR360E
Large Padlock Station Holds up to 4 padlocks 7.6cm 40.6cm
Each for $46.90
Add to cart
 
Description

Description

  • Rugged and eye-catching station
Specifications

Specifications

3.1cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit