Learn More
Brady™ PowerMark™ Indoor/Outdoor Vinyl Tapes
PowerMark™ Vinyl Tapes are made of durable, low-shrink vinyl with ultra-aggressive adhesive that \is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Supplier:  Brady™ 13615
 View more versions of this product
Catalog No. 19-041-128
Add to Cart
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • 8-10 year average outdoor durability in temperatures from -40°F to 180°F (-40°C to 82°C); can be applied as low as 0°F (-18°C)
Specifications
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Gold
Vinyl
Non-sterile
BBP™85, PowerMark™ Printer
8 to 10 Years
-40 to 180°F
0.004 in.
50 ft.
10 in.
Acrylic, Permanent
Tape
Labeling Tape
PowerMark™ label maker
50 Gardner Units
1 Ea.
82 to -40°C
0.1 mm
152 cm
Product Suggestions
Product Suggestions