Learn More
Brady™ Production Status Tags
Production Status Tags help you effectively monitor and control production and equipment maintenance.
Manufacturer:  Brady™ 86740
 View more versions of this product
Catalog No. 19-040-499
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • The excellent write-on surface of B-853 Cardstock is ideal for adding on-the-spot customized notes and other information
  • Paper cardstock tags are recommended for indoor use only
  • Includes 3/8 in. metal grommet
  • Not recommended for chemical exposure
Specifications
Specifications
Black on Yellow
Paper
DESCRIPTION/PART NO./SERIAL NO./JOB NO./P.O. NO.
(100) 12 in. Cotton String Ties