Learn More
Brady™ Production Status Tags
Production Status Tags help you effectively monitor and control production and equipment maintenance.
Manufacturer:  Brady™ 86778
 View more versions of this product
Catalog No. 19-040-856
Add to cart
Description
Description
  • The excellent write-on surface of B-853 Cardstock is ideal for adding on-the-spot customized notes and other information
  • Paper cardstock tags are recommended for indoor use only
  • Includes 3/8 in. metal grommet
  • Not recommended for chemical exposure
Specifications
Specifications
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
9.52 mm
3 x 5.75 in.
Production Status Tag
To help you monitor and control production and equipment maintenance
76.2 mm
−40° to +194°F
0.25 mm
146 mm
0.375 in.
Black on Green
Cardstock
Repaired Equipment I.D./Description/Job No./Serial No./Condition/Comments/By/Date
3 in.
(100) 12 in. cotton string ties
−40° to +90°C
5.75 in.