Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Quick-Align™ Lettering

Quick-Align™ Lettering is featured in Quik-Align™ Numbers & Letters and Instalign™ Numbers & Letters and are perfect for all general identification projects.

$38.25

Specifications

Product Type Hazard Sign
Temperature Range (English) -40°F to 180°F
Certifications/Compliance RoHS
Material Vinyl
Quantity 10/Pk.
View More Specs

Products 2
Catalog Number Mfr. No. Displayed Text Price Quantity & Availability  
Catalog Number Mfr. No. Displayed Text Price Quantity & Availability  
19-016307
View Documents
Brady™
58604
Radiation trefoil
Pack of 10 for $38.25
Add to Cart
 
19-016245
View Documents
Brady™
58509
No Water Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description

Description

  • Card-mounted vinyl film (B-933) characters features a pressure sensitive adhesive, average outdoor durability 2-4 years.
  • Service temperature of -40°F to 200°F (-40°C to 93°C)
  • High gloss, durable, flexible vinyl (B-946)with a permanent cold temperature pressure sensitive acrylic adhesive, average outdoor durability 8-10 years.
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications

Specifications

Hazard Sign
RoHS
10/Pk.
Permanent Acrylic
180°F
0.004 in.
-40°F to 180°F
Vinyl
Gothic
Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
-40°F
-40°C to 82°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit