Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Quik-Align™ Gothic Style Kits, Black

Quik-Align™ die-cut numbers and letters are self-spacing and self-aligning. The characters interlock to ensure perfect spacing for professional-looking results every time.

Specifications

Color Black
Product Type Gothic Quik-Align kit
Displayed Text A thru Z, 0 thru 9, $, & And PUNCTUATION
Temperature Range (English) -40°F to 200°F
Certifications/Compliance ASTM D 2979
View More Specs

Products 5
Catalog Number Mfr. No. Height (English) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Quantity Price Quantity & Availability  
19-016176
View Documents
Brady™
52211
3 in. 1 Kit Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-016179
View Documents
Brady™
52219
6 in. 1 Ea. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-016178
View Documents
Brady™
52215
4 in. 1 Kit Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-108-131
View Documents
Brady™
52203
1 in. 1 Kit Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-108-133
View Documents
Brady™
52207
2 in. 1 Kit Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description

Description

  • Traditional sans serif Gothic font provides excellent legibility even at a distance
  • Card-mounted vinyl film characters features a pressure sensitive adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 2-4 years
Specifications

Specifications

Black
A thru Z, 0 thru 9, $, & And PUNCTUATION
ASTM D 2979
Vinyl
14 lb./in.
Indoor and outdoor use
-40°F
-40°C to 93°C
Gothic Quik-Align kit
-40°F to 200°F
20 Each of A, E, I, L, N, O, R, S, T and numbers 0 to 9 10 each of remaining letters and 10 each of $, & and punctuation
Gothic
Permanent Acrylic
200°F
0.005 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit