Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Red Striped “Do Not Operate” Lockout Tags

Red Striped “Do Not Operate” economy polyester (B-851) lockout tags are perfect for marking on-the-spot warnings of dangerous conditions.

$78.50

Specifications

Certifications/Compliance OSHA 1910.145
Color Black/Red on White
Dimensions (L x W) 5.75 in. H x 3 in. W
Durability 3 to 5 Years
For Use With (Application) Marking on the spot warning of dangerous conditions
View More Specs

Products 1
Catalog Number Mfr. No. Material Displayed Text Price Quantity & Availability  
Catalog Number Mfr. No. Material Displayed Text Price Quantity & Availability  
19-102641
View Documents
Brady™
66063
Economy Polyester (B-851) Danger Front Do Not Operate This lock/tag may only be removed by ETC (with diagonal stripes), Back This energy source has been Locked Out! Unauthorized removal of This lock/tag may result in immediate discharge. Remarks
Pack of 25 for $78.50
Add to cart
 
Description

Description

  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Withstands moisture, grease and dirt
  • Stays legible longer than cardstock
Specifications

Specifications

OSHA 1910.145
5.75 in. H x 3 in. W
Marking on the spot warning of dangerous conditions
146mm
-40° to 100°C
Economy Polyester (B-851)
3 in.
Black/Red on White
3 to 5 Years
5.75 in.
-40° to 212°F
0.25mm
Lockout Tag
76mm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit