Learn More
Brady™ Repair Tags
Repair Tags are perfect for marking on-the-spot warnings of dangerous conditions.
Supplier:  Brady™ 76141
 View more versions of this product
Catalog No. 19-003-337
Description
Description
  • Rigid economy polyester write-on tags meet OSHA Standard 1910.145 for accident prevention.
  • Durable heavy duty write-on polyester (B-851 and B-837) tags with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices.
Specifications
Specifications
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Polyester
General Equipment
14.6cm
7.62 cm
Black, Orange, White
Front: Repair
5.75 in.
3 in.
3 x 5.75 in.