Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Reversible Floor Stands
Reversible Floor Stands are an economical way to get your messages noticed. They are made of lightweight corrugated polypropylene material (B-836).
Supplier: Brady™ 92281
Safety Services Available
Overview
Logo/Customization
Description
- Folds for easy storage when not in use
- Each stand contains two different legends - just turn the stand inside out to display each one
- Front legend: HAZARDOUS AREA (with pictogram)
- Back legend: MEN WORKING (with pictogram)
Specifications
Globally Harmonized System of Classification and Labeling of Chemicals (GHS);NFPA 704;OSHA 1910.1200 | |
Black on Yellow | |
-40°F to 130°F | |
Polypropylene | |
1.5 lb./in. | |
Designed for use in industrial, utility, commercial and institutional environments | |
130°F | |
0.16 in. | |
30.48 cm |
Reversible Floor Stand | |
Front Hazardous Area Back Men Working | |
20 x 12 in. | |
1 Ea. | |
Non-adhesive | |
20 in. | |
-40°F | |
12 in. | |
-40°C to 54°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction