Brady™ Safety Banners Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Brady™ 50907

 View more versions of this product

Catalog No. 19-807-079

  Check Availability
Add to Cart

    Specifications

    Grade Industrial
    Height (English) 3.5 ft.,3.5 ft.
    Material Vinyl
    Message Safety Starts With You!
    Quantity Each
    Thickness 10mil,10mil
    Color Black, Green, Red on White,Black, Green, Red on White
    Type Safety banner
    Width (English) 10 ft.,10 ft.
    For Use With (Application) Indoor use,Indoor use

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit