Learn More
Brady™ Scaffolding Tags
Scaffolding Tags are perfect for marking on-the-spot warnings of dangerous conditions.
Supplier: Brady™ 86683
Description
- Write-on polyester laminate (B-837) protects the graphics, wipes clean and withstands water, grease and extreme temperatures.
- (B-851) Rigid Economy Polyester textured write-on surface allows for authorized signatures, dates and other special instructions.
- The excellent write-on surface of (B-853) Cardstock is ideal for adding on-the-spot customized notes and other information, recommended for indoor use only.
Specifications
ASTM | |
0.37 in. | |
DANGER Do Not Use This Scaffold Keep Off This scaffold is being erected, taken down, or has been found defective. Do Not Alter Inspection Date And Initial | |
5.75 in. | |
100 string fasteners | |
-40°C to 90°C | |
3 in. | |
5.75 x 3 in. |
Black, Red | |
Cardstock | |
Used for temporary tagging in a non harsh environment | |
14.6cm | |
0.01 in. | |
-40°F to 194°F | |
7.6 cm | |
0.93 cm |