Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Self-Laminating Tags
Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.
Supplier: Brady™ 81991
Includes: 10 Nylon ties
Description
- B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
- Textured write-on surface allows for authorized signatures, dates and other special instructions
- Seal and protect your written message with a clear adhesive overlay
- Withstands moisture, grease and dirt
Specifications
10/Pk. | |
Black, Red, White | |
Polyester | |
For marking on-the-spot warning of dangerous conditions | |
7 in. | |
0.01 in. | |
-40°F to 212°F | |
10.1 cm | |
0.93 cm |
OSHA | |
0.37 in. | |
DANGER Do Not Remove This Tag/Remarks/See Other Side | |
3 to 5 Years | |
17.7cm | |
-40°C to 100°C | |
4 in. | |
7 x 4 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction