Learn More
Brady™ Self-Sticking Vinyl Pipe Markers
Self-Sticking Vinyl Pipe Markers from Brady™ simplify installation with their factory-applied industrial-grade adhesives: simply peel off the backing and press the marker in place.
Supplier:  Brady™ 71354
 View more versions of this product
Catalog No. 19-017311
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Banding each end of a pipe marker with directional flow arrow tape
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
  • Style 1 pipe markers measure 2-1/4 in. x 14 in., come one marker per card, and fit pipe diameter 2-1/2 in. to 7-7/8 in.
  • Style 4 pipe markers measure 1-1/8 in. x 7 in., come with four markers per card and fit pipe diameter 3/4 in. to 2-3/8 in.
  • Style 1HV pipe markers measure 4 in. x 24 in., come with one marker per card and fit 8 in. or greater pipe diameter
  • Style 3C pipe markers measure 2-1/4 in. x 2-3/4 in. and fit pipe diameters less than 3/4 in.
Specifications
Specifications
ANSI A13.1
Acrylic, Permanent, Cold Temperature, Pressure-sensitive
High Gloss
Non-sterile
For the identification of piping system contents when used with Directional Flow Arrow Tape
50 Gardner Units
1 Ea.
-40°C to +82°C
1.125 in. H x 7 in. W
177.8 mm
28.575 mm
Pipe Marker
Black, Yellow
Vinyl
High Pressure Air
Pipe & Valve Marking
8 to 10 Years
-40 to 180°F
0.102 mm
7 in.
1.125 in.