Learn More
Brady™ Solid Color Banding Tape
Provide 360° visibility to improve safety and operational efficiency
Manufacturer:  Brady™ 36313
 View more versions of this product
Catalog No. 19-806-995
Add to cart
Description
Description
  • Made of pressure sensitive vinyl
  • Tapes are supplied in liner-mounted rolled form, use two bands per marker
Compliance
Meets ASME color field size recommendation when used in combination with worded legends and directional flow arrows
Specifications
Specifications
Safety
Purple
60 Gardner Units
30 yd.
8 to 10 Years
-40°C to +82°C
2 in.
Vinyl
1 Roll(s)
Acrylic, Pressure-sensitive
ASME (ANSI)
For pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
27.4 m
-40 to 180°F
0.004 in.
Pipe Marker
Pipe Banding Tape