Learn More
Brady™ Solid Color Banding Tape Available on GSA/VA Contract for Federal Government customers only.
Provide 360° visibility to improve safety and operational efficiency
Manufacturer:  Brady™ 91431
 View more versions of this product
Catalog No. 18-999-2582F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Acrylic,Pressure-sensitive
ASME (ANSI)
For pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
27.4m
-40 to 180°F
0.004 in.
Tape
Pipe Banding Tape
White
60 Gardner Units
30 yd.
8 to 10 Years
-40 to 82°C
2 in.
Vinyl
1 Roll(s)