Brady™ SPC Marine Sorbent Booms

Designed to float on water and contain or absorb a wide variety of marine fuels and oils

Manufacturer: Brady™ SPC510

 View more versions of this product

Catalog No. 17-111-090

  Check Availability
Add to Cart

    Specifications

    Capacity (English) Sorbing 32 gal./bale
    For Use With (Application) Outfalls and Discharge Ponds
    Product Type SPC Marine Sorbent Boom
    Dimensions (L x W) 12.7cm x 3.04m (5 in. x 10ft.)
    Packaging Quantity 4/Bale

    • Strong, lint-free, and gapless
    • Class: Oil Only
    • Available in various sizes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit