Learn More
Brady™ SPC Re-Form Oil-Only Heavy-Weight Pads and Rolls Available on GSA/VA Contract for Federal Government customers only.
For drips, spills and wiping applications
Manufacturer:  Brady™ RFOP28DP
 View more versions of this product
Catalog No. 19-102-756
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
268.76L
150 ft.
Chemical resistant sorbent for oil and solvent base liquids, drips, spills and wiping applications
Heavy
72.4cm
150 ft. x 28.5 in. (45.7m x 72.4cm)
71 gal.
45.7m
Re-Form™ Oil Only Sorbent Roll
Re-Form™ Sorbent, Natural Cotton Fiber
28.5 in.
71 gal., 268.76L