Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC Re-Form Universal Sorbent Rolls Available on GSA/VA Contract for Federal Government customers only.

Spunbond coverstock adds durability to handle foot traffic

Manufacturer:  Brady™ RFP14P

 View more versions of this product

Catalog No. 19-190-056


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

NFPA Class A
30.2 gal.
45.7m
Re-Form™ Plus Sorbent Roll
Re-Form™ Plus Sorbent
Heavy
36.19cm
150 ft. x 14.25 in. (45.7m x 36.19cm)
Gray
114.3L
150 ft.
For use in traffic areas, general-purpose use
Coverstock
14.25 in.
30.2 gal., 114.3L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit