Learn More
Brady™ Stainerbondz Super Chemical Resistant Polyester Laboratory Labels
Manufacturer:  Brady™ B33164481
 View more versions of this product
Catalog No. 19-140-688
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Acrylic,Permanent
RoHS
0.9 in.
2.2cm
-80 to 130°C
0.0033 in.
Polyester
2500 Roll(s)
White
Matte
Sample generation, inspection and storage
-112 to 266°F
40 lb./in.
0.9 in.
Laboratory Label