Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Thermal Transfer Printer Labels: Rectangular PROMO

For a limited time, you can three Brady label rolls for the price of two.

Manufacturer:  Brady™ THT8874252PROMO

 View more versions of this product

Catalog No. 11-878-166

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

Acrylic
Matte
19.05mm
Thermal Transfer Printer Labels - 1 in. Core
White
25.4mm
Polypropylene
Pack of 3
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit