Brady™ TLS PC Link™ Thermal Labeling System Available on GSA/VA Contract for Federal Government customers only.
Rugged desktop/portable printer with multiple labeling capabilities
Manufacturer: Brady™ TLSPC
 View more versions of this product
Catalog No. 19-150-100
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Warranty 1 Year(s)
    Includes TLS-PC Link™ printer, AC adapter, Labelmark™ standard, rechargeable battery pack, R6210 ribbon, PTL-19-423 labels, quick start guide, manuals, serial communications cable, Windows™ drivers
    Memory 21000 Characters, 10 label lists
    Power Supply Details Rechargeable battery, AC power (optional), stand alone or PC connectivity via USB
    Quantity Each
    Resolution 203 dpi
    Software Type LabelMark™/CodeSoft™ software
    Certifications/Compliance FCC Class A
    Display LED
    For Use With (Application) Arc flash labeling, barcode labeling, circuit board labeling, data communications labeling, facility, safety and maintenance identification, general and industrial labeling, healthcare identification, laboratory identification, panel identification, product identification, security identification, wire and cable marking
    For Use With (Equipment) Specialty lab materials which are sized to specific vial and tube sizes
    Weight (English) 2.5 lb.
    Weight (Metric) 1.14kg
  • Description & Specifications
Related Products
Product Content Feedback