Learn More
Brady™ TLS 2200?/TLS PC Link? Portable Thermal Transfer Printer Labels for Vial Tops
Labels for vial tops
Manufacturer:  Brady™ PTL82499YL
 View more versions of this product
Catalog No. 11-877-52
  Check Availability
Add to cart
Specifications
Description
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Description & Specifications
Specifications
Acrylic,Permanent
Halogen-free (DIN VDE 0472/parts 815), UL
Surface Mount
Laboratory identification, vial, vial top
-320-194°F
0.3 in.
Nylon Cloth
500 Roll(s)
Yellow
0.9cm
Matte
BMP™71, TLS 2200™, TLS-PC Link™
-196-90°C
Label
TLS 2200™/TLS-PC Link™ Label