Brady™ and TLS-PC Link™Portable Thermal Transfer Printer Labels for Vial Tops

Labels for vial tops

Manufacturer: Brady™ PTL82499

 View more versions of this product

Catalog No. 11-877-72

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Outside 0.3 in.
    Format Label
    Adhesive Type Permanent Acrylic
    Product Type TLS 2200™/TLS-PC Link™ Label
    Style Surface Mount
    Temperature Range (English) -320-194°F
    Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815), UL
    Diameter (Metric) Outside 0.9cm
    Finish Matte
    For Use With (Application) Laboratory Identification, Vial, Vial Top
    For Use With (Equipment) BMP™71, TLS 2200™, TLS-PC Link™
    Material Nylon Cloth
    Quantity 500 Roll(s)
    Color White
    Temperature Range (Metric) -196-90°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit