Learn More
Brady™ and TLS-PC Link™ Tamper-Evident Labels
and TLS-PC Link™ Labels are made of Tamper-Resistant Vinyl (B-351) that is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
Manufacturer:  Brady™ PTL20351
Catalog No. 11-877-45
Add to cart
Description
Description
  • High performance white matte vinyl with permanent adhesive is ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -40 to 180° F (-40 to 82° C)
Specifications
Specifications
Non-sterile
White
RoHS, 2005/618/EC MCV, 2002/95/EC
1 in. H x 2 in. W
Matte
For rating and serial plates that require high performance and resistance to product tampering
2.54cm
Rectangle
63 lb./in.
0.066 mm
2 in.
Label
TLS 2200™/TLS-PC Link™ Label
Acrylic
White
Surface Mount
1 in. H x 2 in. W
1 in.
BMP™71, TLS 2200™, TLS-PC Link™
1
-40 °C-80 °C
0.0028 in
28.57 mm
50.8 mm
Vinyl
100 Roll(s)